Merck
28277-U
Supelco
Siebe für GERSTEL® TD-Röhrchen
greener alternative
pkg of 10 ea
About This Item
UNSPSC-Code:
41104008
NACRES:
NB.24
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
stainless steel
Leistungsmerkmale
(Fits both stainless steel and glass tubes)
Verpackung
pkg of 10 ea
Grünere Alternativprodukt-Eigenschaften
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
Methode(n)
active air sampling: suitable
Anwendung(en)
air monitoring
environmental
industrial hygiene
Kompatibilität
for use with Gerstel®
Grünere Alternativprodukt-Kategorie
Allgemeine Beschreibung
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.
Rechtliche Hinweise
GERSTEL is a registered trademark of Gerstel GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)