Merck
27081-U
Supelco
Fläschchen, Schraubkappe, Klarglas
marking spot
, volume 2 mL, O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm, thread for 8-425, pkg of 1000 ea
About This Item
UNSPSC-Code:
41122001
NACRES:
NB.11
Materialien
clear glass vial (with marking spot)
Beschreibung
cap not included
Leistungsmerkmale
closure type screw top vial
Verpackung
pkg of 1000 ea
AD × H × ID
12 mm × 32 mm × 4.6 mm
Volumen
2 mL
Fitting
thread for 8-425
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)