2630905
Inlet-Liner, Splitless-Typ, gerades Design (ungepackt)
pkg of 5 ea
Verpackung
pkg of 5 ea
L × AD × ID
92 mm × 6.35 mm × 4.0 mm
Allgemeine Beschreibung
- Length 92 mm × O.D. 0.25 in. × I.D. 4 mm
- Application: General purpose
- Mfr. Part No.: N610-1052, N612-1001
Choose from one of the most recent versions:
Analysenzertifikate (COA)