Merck
23166
Supelco
Quetschpumpe 2 ml
About This Item
UNSPSC-Code:
41112500
Verpackung
pkg of 10 ea
Allgemeine Beschreibung
Squeeze bulb is a hand operated sampler which is suitable for obtaining small grab samples. Squeezing the bulb ejects air through one way valve. They could be attached to the sample compartments of gas chromatographs.[1]
Anwendung
Squeeze bulb may be used to force petroleum ether out of the column during HPLC analysis to determine the concentration of surfactants from sample with XAD-2 resin.[2]
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
HPLC determination of ethoxylated alcohol surfactants in wastewater.
Schmitt, Th M., et al.
Journal of the American Oil Chemists' Society, 67, 103-109 (1990)
J. R. Mudakavi
Principles and Practices of Air Pollution Control and Analysis, 145-145 (2010)