Merck
22117
Feindosierventil
stainless steel, 1/8 in. straight arms
Synonym(e):
Swagelok® SS2,SS FEINDOSIERVENTIL 1/8S, 1 STÜCK
About This Item
UNSPSC-Code:
41115717
eCl@ss:
32035302
Materialien
stainless steel
Fitting
1/8 in. straight arms
Allgemeine Beschreibung
Für die exakte Einstellung des Gasflusses.
Mit Nuts und Ferrules.
Rechtliche Hinweise
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Analysenzertifikate (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Kundensupport
Die Dokumentenbibliothek aufrufen
Critical micelle concentrations for alkyltrimethylammonium bromides in water from 25 to 160 C.
Fennell ED
Journal of Solution Chemistry, 13 (2), 87-101 (1984)