Merck
Z757764
Sartorius filter pipette tips
volume range 0.1-10 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Synonym(e):
filter pipette tips, pipetting tips, pipettor tips
Größe auswählen
960 EA
186,00 €
186,00 €
Versandbereit am11. April 2025Details
Größe auswählen
Ansicht ändern
960 EA
186,00 €
About This Item
UNSPSC-Code:
41121601
NACRES:
NB.01
186,00 €
Versandbereit am11. April 2025Details
Materialien
colorless
polypropylene (Virgin)
Sterilität
sterile
Leistungsmerkmale
DNase free
RNase free
barrier
filter
Verpackung
pack of 960 ea (10 racks of 96 tips)
Hersteller/Markenname
Sartorius 790011F
Maximales Volumen
10 μL
Volumenbereich
0.1-10 μL
Volumen
0.1—10 μL
Bindungsart
low binding surface
Allgemeine Beschreibung
The Sartorius filter tips of the Biohitfamily are made of polypropylene and feature a polyethylene filter barrier to prevent aerosol and liquid contamination. This filter helps protect against the rist of cross-contamination and reduces maintenance requirements of the pipette. The filter tips are suited for molecular biology, microbiology, cell culture and radioactive work.
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
  • Pipette foaming liquids like buffers and proteins
  • Pipette solvents
  • Reverse pipetting
  • Multiple dispensing functions of electronic pipettes
Sonstige Hinweise
Browse all pipette tips
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)