Merck
Z756903
Mouse restrainer with dorsal access
For mice 18-35 grams, pack of 1 ea
Synonym(e):
Rodent restrainer
Größe auswählen
1 EA
92,60 €
92,60 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
92,60 €
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.22
92,60 €
Check Cart for Availability
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F46401-0000
Allgemeine Beschreibung
The accurate, safe and fast way to give injections. The dorsal skin of the mouse is easily accessed through the top circular opening so you can accurately perform subcutaneous injections without risk of being bitten and less likelihood of sticking your own finger.
  • Easy entry: draw mouse tail first or lead head first through the wide entry funnel into the back of the tube and then position stop gate in choice of two slots.
  • With tail first entry, tail is exposed at rear of tube allowing easy access for tattooing and tail bleeds
  • Overall Dimensions (L x W x H): 17.4 x 10 x 6.7cm (6.8 x 3.9 x 2.6 in.)
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)