Merck
Z741754
Manifold
2-Place
Größe auswählen
1 EA
73,20 €
73,20 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
73,20 €
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.22
73,20 €
Check Cart for Availability
Materialien
polysulfone
Leistungsmerkmale
autoclavable
Includes necessary tubing – 1/8” i.d. silicone
Hersteller/Markenname
IBI ACCFL0005
Allgemeine Beschreibung
The manifold allows for the connection and simultaneous delivery of media to flow cells and for consolidation of effluent from cells to the waste bottle. Comes with 1/8” i.d. silicone tubing attached and is autoclavable and re-usable.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)