Merck
Z741658
Pipetting Reagent Vessel
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Materialien
polypropylene
Verpackung
pack of 16
Hersteller/Markenname
Sartorius 783500
Kapazität
120 mL
Allgemeine Beschreibung
Made of polypropylene, the autoclavable and durable reagent vessel is chemically resistant to all common reagents. It is especially useful with all multichannel pipettes.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)