Merck
Z692565
microDOC Basic gel documentation system
AC/DC input 230 V AC
Größe auswählen
1 EA
2.600,00 €
2.600,00 €
Voraussichtliches Versanddatum29. Mai 2025
Größe auswählen
Ansicht ändern
1 EA
2.600,00 €
About This Item
UNSPSC-Code:
41105300
NACRES:
NB.22
2.600,00 €
Voraussichtliches Versanddatum29. Mai 2025
Leistungsmerkmale
Contains UK and Schuko Plugs
Hersteller/Markenname
Cleaver CSL-MDOCBASIC
Allgemeine Beschreibung
System contains: Canon G12 digital camera, basic lift-off dark room hood.
This is the perfect low cost option to upgrade to digital documentation of your gels. Can be used with most 21 x 21 cm tranilluminators.
Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
  • Canon G15 digital camera (12.1 MP)
  • 2GB storage card
  • Ethidium bromide & close up +3 filters
  • Lift-off hood
  • Available with optional TotalLab 1D software
Rechtliche Hinweise
TotalLab is a trademark of Nonlinear Dynamics Ltd.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)