Merck
Z659711
Bandelin SONOREX Ultrasonic bath accessories
positioning lid for Z659673 and Z659029
Größe auswählen
1 EA
61,00 €
61,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
61,00 €
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.74
61,00 €
Check Cart for Availability
Materialien
stainless steel
Leistungsmerkmale
for 2 beakers
Verpackung
pack of 1 ea
Hersteller/Markenname
Bandelin DE 255
Verfügbarkeit
not available in USA and Canada
Allgemeine Beschreibung
When using chemically aggressive liquids, removing chemically aggressive or abrasive contamination or sonicating sample liquids, insert cups are used (indirect sonication). Because nothing may be placed on the bottom of the ultrasonic bath, use a suitable perforated lids/insert strips to hold the insert cups. Several insert cups can be sonicated with different liquids at the same time.
Leistungsmerkmale und Vorteile
  • for holding insert cups
  • sound-absorbing
  • shortens the heating phase
Rechtliche Hinweise
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)