Merck
Z373443
Sigma-Aldrich
LiposoFast Liposome Factory
Replacement gas-tight 0.5 mL syringe
Größe auswählen
1 EA
152,00 €
152,00 €
Versandbereit am02. April 2025Details
Größe auswählen
Ansicht ändern
1 EA
152,00 €
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.22
152,00 €
Versandbereit am02. April 2025Details
Allgemeine Beschreibung
Fast, efficient formation of uniform-sized, unilamellar liposomes. The principle is simple: a lipid emulsion is repeatedly extruded through a porous polycarbonate membrane, forced back and forth by specially modified gas-tight syringes attached to the membrane support capsule. With the syringes provided, unit has 0.5 mL capacity and virtually zero dead volume. Membranes from 100 to 400 nm pore size available.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)