Merck
S4642
Wheaton staining dishes
for 20 slides, dishes only
Synonym(e):
900201
Größe auswählen
3 EA
165,00 €
165,00 €
Auf LagerDetails
Größe auswählen
Ansicht ändern
3 EA
165,00 €
About This Item
UNSPSC-Code:
41121812
NACRES:
MA.01
165,00 €
Auf LagerDetails
Materialien
soda-lime glass
Verpackung
pack of 3 ea
Hersteller/Markenname
WHEATON® 900201
Schalenkapazität
3 x 1 in. (75 x 25 mm) , 10 single slides
3 x 1-1/2 in. (75 x 38 mm) , 19 slides arranged alternately straight across and diagonally
3 x 2 in. (75 x 51 mm) , 20 slides back-to-back of standard size
inside L × W × D
95 mm × 76 mm × 64 mm
Allgemeine Beschreibung
Microscopy is a method of viewing objects that lie outside the resolution of the human eye and can be examined. The objects to be observed are magnified to such an extent that they can be displayed as images. For sample preparation, the objects to be examined are cut into very thin slices. These sections are then applied to an object slide and covered with a cover glass and appropriate solutions. To view the sections under the microscope, the sections on the slide must be filled in a staining trough and stained with an appropriate staining solution.
Rechtliche Hinweise
WHEATON is a registered trademark of Wheaton Industries, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)