Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | 81,00 € |
Über diesen Artikel
81,00 €
material
acrylic , clear (bin)
feature
holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS23401
L × W × H
184 mm × 194 mm × 232 mm
General description
Features and Benefits
- One Compartment
- See inventory levels of pipette tips, tubes, fittings, finger cots, and other bulk small items at a glance
- Flip-top covers on generous openings make filling and dispensing easy and keep products dust-free
- Includes non-skid rubber feet
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description clear (bin) | description clear (bin) | description clear (bin) | description clear (bin) |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| material acrylic , clear (bin) | material acrylic , clear (bin) | material clear (bin), acrylic | material acrylic , clear (bin) |
| manufacturer/tradename Heathrow Scientific HS23401 | manufacturer/tradename Heathrow Scientific HS23402 | manufacturer/tradename Heathrow Scientific HS23403 | manufacturer/tradename Heathrow Scientific HS23404 |
| L × W × H 184 mm × 194 mm × 232 mm | L × W × H 150 mm × 177 mm × 368 mm | L × W × H 217 mm × 170 mm × 217 mm | L × W × H 287 mm × 170 mm × 217 mm |
| feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories | feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories | feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories | feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?