Merck
GE28-9068-43
Hyperfilm MP
W × L 18 cm × 24 cm, pack of 50 ea
About This Item
UNSPSC-Code:
41105300
NACRES:
NB.22
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sterilität
non-sterile
Verpackung
pack of 50 ea
Hersteller/Markenname
Cytiva 28906843
B × L
18 cm × 24 cm
Verwandte Kategorien
Allgemeine Beschreibung
Amersham Hyperfilm MP is an excellent multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer. The anti-scratch coating reduces the risk of visible artifacts (e.g., plastic wrap marks) and the clear colorless base results in easy reading of images, without compromising on sensitivity.
Leistungsmerkmale und Vorteile
  • High sensitivity for low concentration targets.
  • Intended for radiometric or chemiluminescent applications.
  • Clear background for excellent contrast and improved band visibility.
  • Publication-quality images.
  • High resolution at an economical price.
  • Easy to use.
Lagerung und Haltbarkeit
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Rechtliche Hinweise
Hyperfilm is a trademark of Cytiva
Lagerklassenschlüssel
13 - Non Combustible Solids
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)