Merck
GBL204878
GRACE BIO-LABS SEALSTRIPS AND SEALSTRIP APPLICATOR, FOR PROPLATE® MICROARRAY SYSTEMS
for ProPlate® microarray systems, pack of 1
Synonym(e):
microarray system applicator, seal strip applicator
Größe auswählen
1 EA
23,50 €
23,50 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
23,50 €
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.17
23,50 €
Check Cart for Availability
Leistungsmerkmale
for ProPlate® microarray systems
Verpackung
pack of 1
Hersteller/Markenname
Grace Bio-Labs 204878
Allgemeine Beschreibung
This plastic applicator helps to apply seal strips to the top surface of the chambers. Seal strips are ideal to prevent evaporation, they remove cleanly and easily without leaving a residue.
Rechtliche Hinweise
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
SealStrips is a trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)