Merck
EPM1192-9901
Quick-change platform kit for Eppendorf® New Brunswick Innova® 2100, 4000, 4080, 4230 shaker
enables platform attachment without tools
Synonym(e):
Quick Change Platform Kit
About This Item
UNSPSC-Code:
41103816
NACRES:
NB.73
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Leistungsmerkmale
enables platform attachment without tools
Hersteller/Markenname
Eppendorf® M1192-9901
Verwandte Kategorien
Allgemeine Beschreibung
The Eppendorf M1192-9901 quick-change platform accessory kit converts Eppendorf shakers to allow interchangeable platforms to be installed without tools. It is for use with the Eppendorf New Brunswick Innova 2100 series platform shaker, Excella E24/E24R series incubator shaker, and Innova 40/40R and 42/42R series incubator shaker (all sold separately). The kit includes a subplatform with spring clips, counterweight, and installation hardware.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)