EP6731070166
Eppendorf® Door Lock For CellXpert CO2 Incubator
retrofittable
Synonym(e):
Eppendorf® Door Lock For CellXpert CO2 Incubator, Incubators
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
Empfohlene Produkte
Verpackung
pkg of 1 ea
Hersteller/Markenname
Eppendorf® 6731070166
Allgemeine Beschreibung
Door lock to fit CellXpert C170i CO2 incubators. Fully retrofittable by end user and requires no service engineer.
Leistungsmerkmale und Vorteile
- Retrofittable to all CellXpert C170i produced after January 2022 (Serial number > 6731LH322982)
- No Eppendorf Service required for installation
- No control by user management system or open/close status tracking
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)