Merck
EP5353040148
Tube Holders for Eppendorf® MixMate
new
to hold, 8 x 5-15 mL tubes
Synonym(e):
Racks
About This Item
UNSPSC-Code:
41121800
Leistungsmerkmale
to hold 8 x 5-15 mL tubes
Verpackung
pkg of 1 ea
Hersteller/Markenname
Eppendorf® 5353040148
Allgemeine Beschreibung
Tube Holder 5/15 mL, for 8 x 5/15 mL conical tubes. For use with Eppendorf MixMate.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG