Merck
EP022662970
Lid for Eppendorf(R) F-45-30-11 Rotor
non aerosol tight, for 5804/5804R & 5810/5810R, aluminum
About This Item
UNSPSC-Code:
41103913
Materialien
aluminum
Leistungsmerkmale
non aerosol tight
Hersteller/Markenname
Eppendorf® 022662970 (NA)
Methode(n)
centrifugation: suitable
Kompatibilität
for use with 5804 R
for use with 5804
for use with 5810 R
for use with 5810
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG