Merck
EP0030132513
Eppendorf® Twin tec real-time PCR plates
white plate, skirted, pkg of 25 ea
Größe auswählen
25 EA
196,00 €
196,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
25 EA
196,00 €
About This Item
UNSPSC-Code:
41106300
NACRES:
NB.15
196,00 €
Check Cart for Availability
Materialien
conical bottom white wells
white plate
Leistungsmerkmale
skirt
Verpackung
pkg of 25 ea
Hersteller/Markenname
Eppendorf® 0030132513
Methode(n)
PCR: suitable
Größe
96 well
Arbeitsvolumen
150 μL
Allgemeine Beschreibung
The limiting factor in low-volume real-time PCR (qPCR) is often the remaining fluorescence intensity. Eppendorf twin.tec real-time PCR plates are specially manufactured for qPCR, as they offer all the advantages of regular twin.tec plates, but with the added advantage of white wells. These reflect fluorescence much better than clear or frosted wells, so lower levels of fluorescence are still detectable with the same instrument – just by using the right consumables. The qPCR plates′ white wells also significantly reduce interfering background fluorescence and lead to increased consistency between replicates, and giving highly reproducible results. The wells are made from polypropylene – which guarantees fast heat transfer for fast assays – and the frame is made of rigid polycarbonate, to ensure easy and reliable handling – both in manual and automated assays.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)