EP0030106318
Eppendorf® UVette® routine pack, reclosable box
pkg of 200 ea
Synonym(e):
Staining, Staining Accessories
Größe auswählen
146,00 €
Versandbereit am24. April 2025Details
Größe auswählen
About This Item
146,00 €
Versandbereit am24. April 2025Details
Empfohlene Produkte
Verpackung
pkg of 200 ea
Hersteller/Markenname
Eppendorf® 0030106318
Leistungsmerkmale und Vorteile
- UVette® in routine packs are ideal for routine applications.
- These packs allow a safe storage of the UVettes in a reclosable box, while offering convenient access to each cuvette for easy filling.
- UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons.
- Two built-in optical path lengths in a single cuvette - turn the UVette by 90° to change from 10 mm and 2 mm.
- Volume markings for 500 μL and 1,000 μL.
- Deep-lying optical window prevents scratches.
- Tapered cuvette base for optimal filling, frosted grip for labeling.
- Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers.
- Routine pack for convenient access to each UVette and safe storage in a reclosable box.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)