CLS3081150CO
Pyrex® small knob top desiccator
cover only
Synonym(e):
equipment replacement accessory
Alle Fotos(1)
About This Item
UNSPSC-Code:
41104510
NACRES:
NB.43
Empfohlene Produkte
Materialien
borosilicate glass
Hersteller/Markenname
Corning 3081-150CO
Allgemeine Beschreibung
This 2.4 L Pyrex desiccator is designed for general analytical work. The sturdy knob makes the cover easy to handle.
Warning: Do not heat or subject to pressure. Not for vacuum applications.
Warning: Do not heat or subject to pressure. Not for vacuum applications.
This is the desiccator cover replacement which fits Pyrex desiccator CLS3181150.
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)