Merck
Z758876
Bollé strap conversion kit for Cobra safety spectacles
for ANSI compliant Cobra spectacles
Synonym(e):
protective eyewear, protective glasses, safety eyeglasses, safety eyewear, safety glasses, safety spectacles
About This Item
UNSPSC-Code:
46181802
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Leistungsmerkmale
for ANSI compliant Cobra spectacles
Verpackung
pkg of 1 ea
Hersteller/Markenname
Bollé 40043
Allgemeine Beschreibung
These optional strap conversion kits for the Cobra safety spectacles allow the ability to interchange the temples with an adjustable strap and foam option. Use these kits with the Cobra wrap around safety spectacles, sold separately: Z758833 or Z758841
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)