Merck
Z743201
Julabo® bath carrier trays for SW22/SW23
basic tray for assembling spring clamps
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 8970620
Allgemeine Beschreibung
Basic stainless steel tray for assembling spring clamps. For use with Julabo® shaking water bath models SW22, SW23. Mix and match combinations of spring clamps Z743249 (8970601) through Z557471 (8970609) install on the basic tray.
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)