Z743027
Julabo® CORIO™ CD heating circulator with open bath
CD-B39, capacity 35 L (41 L), temperature 20-150 °C, stainless steel bath, AC/DC input 230 V AC, UK plug
Größe auswählen
3.300,00 €
Größe auswählen
About This Item
3.300,00 €
Empfohlene Produkte
Materialien
stainless steel bath
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 9012439
Temperaturmessbereich
20-150 °C
Kapazität
35 L (41 L)
Außenmaß B × L × H
54 cm × 34 cm × 57 cm
Öffnung B × L × H
33 cm × 30 cm × 30 cm
Allgemeine Beschreibung
Your advantages
- Models for internal and external applications
- Bright, white, easy to read display
- Very quiet
- Easy pump change-over between internal and external circulation
- USB connection
- High-quality bath tanks made of stainless steel
- Integrated drain screw (except B5)
Cooling coil and bath cover available as optional accessories
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)