Merck
Z742911
Kern FCB series bench scale
Kern FCB 12K1, weighing capacity 12000 g, accuracy: 1.0 g, precision: ±3.0 g
Größe auswählen
1 EA
230,00 €
230,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
230,00 €
About This Item
UNSPSC-Code:
41111502
NACRES:
NB.71
230,00 €
Check Cart for Availability
Aufnahme AC/DC
100 - 240 V AC, universal plug set
Hersteller/Markenname
Kern FCB 12K1
Messbereich Wiegekapazität
12000 g, accuracy: 1.0 g, precision: ±3.0 g
Allgemeine Beschreibung
Bench scale with second display on the backside of the scale.
PRE-TARE function for manual subtraction of a known container weight, useful for checking fill-levels
High mobility: Battery operation option, compact construction and low weight make this balance ideal for use in several locations (production, warehouse, dispatch department, ...)
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)