Merck
Z742868
Kern PLS series precision balances
Kern PLS 720-3A, weighing capacity 720 g, accuracy: 0.001 g, precision: ±0.002 g
Synonym(e):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
Größe auswählen
1 EA
1.170,00 €
1.170,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
1.170,00 €
About This Item
UNSPSC-Code:
41111502
NACRES:
NB.71
1.170,00 €
Check Cart for Availability
Aufnahme AC/DC
110 - 240 V AC, universal plug set
Verpackung
pack of 1 ea
Hersteller/Markenname
Kern PLS 720-3A
Messbereich Wiegekapazität
720 g, accuracy: 0.001 g, precision: ±0.002 g
Allgemeine Beschreibung
Kern precision balances offer maximum accuracy for demanding measuring tasks in laboratories, industry and research. With versatile functions, simple operation and robust construction, they are the ideal solution for reliable results in any environment. The PLS series of balances offers a high-quality precision scale with comfortable graphic display and enormous weighing range.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)