Z693545
2mag bioMIXcontrol MS4 control unit
AC/DC input 230 V AC, UK plug
Synonym(e):
laboratory stirrers, magnetic stir plates, magnetic stirrers, stirrers
Größe auswählen
2.730,00 €
Größe auswählen
About This Item
2.730,00 €
Empfohlene Produkte
Materialien
stainless steel plate
Leistungsmerkmale
CE compliant
Verpackung
pack of 1 ea
Hersteller/Markenname
2mag 98604
Parameter
5 to 250 rpm speed (stirring)
5-250 rpm speed
Plattengröße × B × L
× 200 mm × 110 mm
Kapazität
capacity (4 x bioMIXdrive 1)
Allgemeine Beschreibung
Control unit for the connection of 4 stirring drive bioMIXdrive 1, individual and independent speed setting for each stirring drive (Master-Slave-Technology), also ON/OFF function, synchronous speed mode for all drives, extended speed range from 5 to 250 rpm, 6 watts/stirring point without any own-generated heating effect, automatic power setting for the warming-free continuous operation e.g. in CO2-incubators, jerk-free stirring, even and smooth stirring also at slow speeds.
Digital display for speed setting, SoftStart for reliable and safe accelerating of stirring bar, QuickSet for quick setting of start- and maximum speed, storage of last working status when turning off, firm stainless steel housing, easy to clean.
Interfaces: 0-10 volts, 4-20 mA, RS 232, USB, Ethernet.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)