Merck
Z630004
IKA® A 11.2 basic grinding mill cutting blade
A 11.2 cutting blade, 1/cs
Größe auswählen
1 EA
215,00 €
215,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
215,00 €
About This Item
UNSPSC-Code:
41101708
NACRES:
NB.74
215,00 €
Check Cart for Availability
Materialien
stainless steel
Verpackung
pack of 1
case of 1 ×
Hersteller/Markenname
IKA 2905200
Eignung
suitable for (pulverizing soft, fibrous grinding materials.)
Allgemeine Beschreibung
Cutting blade for use with the IKA A11 Basic Analytical Mill, for cutting soft, fibrous ground material. Made of stainless steel
Rechtliche Hinweise
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)