Merck
Z569631
KDS syringe multi-rack for Legato series pumps
large
Synonym(e):
dosing pump syringe rack, infusion pump syringe rack
Größe auswählen
1 EA
1.040,00 €
1.040,00 €
Voraussichtliches Versanddatum17. April 2025
Größe auswählen
Ansicht ändern
1 EA
1.040,00 €
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.01
1.040,00 €
Voraussichtliches Versanddatum17. April 2025
Verpackung
pack of 1 ea
Hersteller/Markenname
KD Scientific 78-8301
Allgemeine Beschreibung
Accommodates up to 4 x 60 to 140 mL syringes. Works with Legato 200, 210 and 210P.
Rechtliche Hinweise
Legato is a trademark of KD Scientific
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Fragen
  1. What is the lowest possible flow rate of Z569631 KDS syringe multi-rack for Legato™ series pumps
    1 Antwort
    1. The minimum flow rate is dependent upon the chosen syringe size. This product has a minimum flow rate of 5 pl/min with a 0.5 µl syringe. The maximum flow rate is 215.803 ml/min with a 140 ml syringe. The graphic below offers additional minimum and maximum flow rates by syringe type.
      Hilfreich?