Z568708
Aldrich® combustion apparatus platinum sample carrier
pkg of 1 ea
Synonym(e):
Combustion Apparatus
Verpackung
pkg of 1 ea
Allgemeine Beschreibung
Has a basket on lower end that holds the paper-wrapped sample.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC