Merck
Z567191
Ace round-bottom pressure flask
with Ace-Thred 15 PTFE front-seal plug, capacity 250 mL
Synonym(e):
Round-Bottom Flask
Größe auswählen
1 EA
166,00 €
166,00 €
Versandbereit am28. April 2025Details
Größe auswählen
Ansicht ändern
1 EA
166,00 €
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
166,00 €
Versandbereit am28. April 2025Details
Materialien
borosilicate glass
round bottom flask
single-neck
Sterilität
non-sterile
Leistungsmerkmale
closure type PTFE plug seal
Verpackung
pack of 1
Hersteller/Markenname
Ace Glass 841511
Kapazität
250 mL
Durchmesser
82 mm
Allgemeine Beschreibung
Heavy-wall borosilicate glass flasks are pressure rated to 60 psig at 120°C. The rounded bottom facilitates use in heating mantles and for other round-bottom flask applications. Flasks are supplied complete with Ace-Thred PTFE front seal bushing and FETFE® O-ring.
Replacement FETFE O-rings:
Ace-Thred #Product #
7 Z504661
15 Z504858
25 Z505056
14 Z504742
210 Z505021
220 Z505110
CAUTION: This glassware is not guaranteed against breakage caused by pressure or vacuum. These flasks should never be used if scratched or otherwise damaged.
Rechtliche Hinweise
FETFE is a registered trademark of Ace Glass, Inc.
Ähnliches Produkt
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Fragen
  1. What could be recommended O-ring to use (with diameter) with this ace RBF? I need an O-ring resistive towards polar and non-polar solvents and should be inert towards chemicals reactions?
    1 Antwort
    1. This is a product of Ace Glass. The plug and O-ring are included. The O-ring for the 250ml flask is product Z504858, which is composed of FETFE and has an I.D. 9.2 mm, and a wall size of 2.6 mm. The general compatibility is listed as 'good', however, it would be recommended to review each component. See the link below to review the Ace Glass O-ring compatibility guide.
      Ace O-Ring Compatibility Guide
      https://www.aceglass.com/downloads/eccc.pdf
      Hilfreich?