Merck
Z531839
Sigma-Aldrich
Tubing adapter with Torion connection and stopcock
O.D. 3/16 in., PTFE stopcock, male joint: ST/NS 14/20
Größe auswählen
1 EA
211,00 €
211,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
211,00 €
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
211,00 €
Check Cart for Availability
Materialien
PTFE stopcock
Verbindung
male joint: ST/NS14/20
AD
3/16 in.
4.8 mm
Allgemeine Beschreibung
PTFE stopcocks are threaded for additional safety and have a 2 mm bore for 3/16 in. tubing connection and 4 mm bore for 5/16 in. and 1/2 in. tubing connections. 5/16in. O.D fits a #8 Torion, and 1/2in. O.D. fits a #14 Torion.
Rechtliche Hinweise
Torion is a trademark of Bibby Sterlin Ltd.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)