Merck
Z510823
Sigma-Aldrich
Connecting Adapter
female joint: ST/NS 19/22, thread size 20-400
Synonym(e):
747130-2019
Größe auswählen
1 EA
48,60 €
48,60 €
Auf LagerDetails
Größe auswählen
Ansicht ändern
1 EA
48,60 €
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
48,60 €
Auf LagerDetails
Gewindegröße
20-400
Verbindung
female joint: ST/NS19/22
Allgemeine Beschreibung
Connects ground joint glassware to threaded microscale component or sample vials. Excellent for use with splash-guards to recover small quantities of material during rotary evaporation. Order threaded vial connectors separately.
Leistungsmerkmale und Vorteile
• Accessory for microscale MICROFLEX threaded kits• Connects Standard Taper outer ground joint to GPI thread of inner ground joint• Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)