Merck
Z341797
IKA® A11 basic grinding chambers
A 11.4 grinding chamber, 1/cs
Synonym(e):
grinders, grinding chambers, grinding mills
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.74
Leistungsmerkmale
CE compliant
CSA approved
Verpackung
case of 1
pack of 1
Hersteller/Markenname
IKA 2904100
Verfügbarkeit
not available in USA and Canada
Größe
250 mL , effective volume
Allgemeine Beschreibung
Note: Made of polycarbonate with stainless steel inlet. Not suitable for cooling with N2, only applicable with double beater A 11.6.
Rechtliche Hinweise
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)