Merck
Z300926
Quickfit® Reduktionsadapter
top female joint: ST/NS 19/26, bottom male joint: ST/NS 29/32
Synonym(e):
DA24
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Verbindung
female joint: ST/NS top19/26
male joint: ST/NS bottom29/32
Hersteller/Markenname
Quickfit DA 24
Verfügbarkeit
not available in USA and Canada
Allgemeine Beschreibung
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Rechtliche Hinweise
Quickfit is a registered trademark of SciLabware Limited