Merck
WHA2810266
Whatman® Hochlelistungs-Extraktionshülsen aus Cellulose
double thickness, L × W 26 mm × 60 mm
Synonym(e):
Z753513
About This Item
UNSPSC-Code:
41104817
NACRES:
NB.43
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Verpackung
pk of 25
Hersteller/Markenname
Whatman 2810-266
Whatman Article No. 28419008 (US reference)
L × B
26 mm × 60 mm
Verwandte Kategorien
Allgemeine Beschreibung
Cytiva′s Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Used in air and waste gas analysis.
Cellulose thimble options for Soxhlet extractor systems
Standard extraction thimbles have a wall thickness of 1–1.5 mm. Double-thickness thimbles have a wall thickness of approximately 2 mm for applications that require higher retention, increased wet or dry strength, or increased rigidity. Measurements can be matched to specific Soxhlet extractor systems.
Leistungsmerkmale und Vorteile
  • Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
  • Suitable for temperatures up to 120°C (248°F)
  • Excellent mechanical strength and retention
  • Wall thickness of standard thimbles is 1–1.5 mm to support application need
  • Option for double thickness for added rigidity
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)