Merck
WHA10441201
Whatman® glass vacuum filtration device
GV025 series, GV025/2/02 hose connector RD14
Synonym(e):
Z750816
About This Item
UNSPSC-Code:
41104900
NACRES:
NB.24
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
borosilicate glass funnel(s) (funnel)
stainless steel clamp(s) (clamp)
Verpackung
pk of 1
Hersteller/Markenname
Whatman 10441201
Whatman Article No. 28415549 (US reference)
Eignung
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
Allgemeine Beschreibung
Whatman® Vacuum Type Glass Holders
Produced from borosilicate glass and available with a choice of support screen. Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
The sintered glass support is recommended for filtration and biological analysis. The 304 stainless steel support screen is suitable for use with proteinaceous solutions.
Leistungsmerkmale und Vorteile
  • Accessory to GV025 Glass Filter Funnel
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)