Merck
SYNM522405
Synthware vacuum manifold with high vacuum valves
number of ports, 5, Hose Connection: left
Synonym(e):
Synthware M522405, vacuum manifold, gas manifold
Größe auswählen
1 EA
614,00 €
614,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
614,00 €
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
614,00 €
Check Cart for Availability
Leistungsmerkmale
number of ports 5
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M522405
Allgemeine Beschreibung
With a serrated hose connection(10 mm O.D.) at left hand side. Main tube is 22 mm O.D. and the distance between valves is 100 mm center to center.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)