SYNF341125
Synthware™ two neck pear shaped flask
25 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonym(e):
Synthware F341125, 2-neck pear shape flask
About This Item
Empfohlene Produkte
Materialien
borosilicate glass flask
pear-shaped
two-neck (angled side neck)
Verbindung
center joint: ST/NS14/20
side joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F341125
Kapazität
25 mL
25 mL