Merck
SYNC501120
Synthware Liebig condenser with 8 mm OD removable hose connections
joint: ST/NS 14/20, jacket L 120 mm
Synonym(e):
Synthware C501120, straight condenser
Größe auswählen
1 EA
101,00 €
101,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
101,00 €
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
101,00 €
Check Cart for Availability
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware C501120
Mantel L
120 mm
Verwandte Kategorien
Allgemeine Beschreibung
Synthware Liebig condenser features a 120 mm jacketed length for superior cooling efficiency. With standard taper outer joint at the top (14/20) and a lower inner drip tip joint. Made from durable borosilicate glass.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)