SYNC179110
Synthware™ water jacketed distillation column with indents to support packing material
joint: ST/NS 19/22, jacket H 110 mm
Synonym(e):
Synthware C179110, jacketed glass distilling column
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware C179110
Mantel H
110 mm
Verwandte Kategorien
Allgemeine Beschreibung
Indentations at the bottom for supporting packing material. Jacket permits the column to be used as a condenser.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)