Merck
SYNA282914
Synthware anti-splash adapter
100 mL, top joint: ST/NS 29/42, bottom joint: ST/NS 14/20
Synonym(e):
Synthware A282914, anti-splash adapter, distillation adapter, adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
bottom joint: ST/NS14/20
joint: ST/NS top29/42
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A282914
Kapazität
100 mL
Verwandte Kategorien
Allgemeine Beschreibung
This design prevents possible contamination from rotary evaporator.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.