Merck
R497029
7-HEXADECYL-8-{(2Z)-2-[1-(4-METHOXYPHENYL)ETHYLIDENE]HYDRAZINO}-3-METHYL-3,7-DIHYDRO-1H-PURINE-2,6-DIONE
AldrichCPR
Größe auswählen
10MG
29,10 €
29,10 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
10MG
29,10 €
About This Item
Lineare Formel:
C31H48N6O3
CAS-Nummer:
Molekulargewicht:
552.767
MDL-Nummer:
29,10 €
Check Cart for Availability
Sonstige Hinweise
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)