Merck
R431249
2-PHENOXYETHYL 3A,8B-DIHYDROXY-2-METHYL-4-OXO-1,3A,4,8B-TETRAHYDROINDENO[1,2-B]PYRROLE-3-CARBOXYLATE
AldrichCPR
Größe auswählen
25MG
29,10 €
29,10 €
Voraussichtliches Versanddatum24. April 2025
Größe auswählen
Ansicht ändern
25MG
29,10 €
About This Item
Lineare Formel:
C21H19NO6
CAS-Nummer:
Molekulargewicht:
381.389
MDL-Nummer:
29,10 €
Voraussichtliches Versanddatum24. April 2025
Sonstige Hinweise
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)