Merck
R386170
(3-(4-ETHOXY-PHENOXY)-2-METHYL-4-OXO-4H-CHROMEN-7-YLOXY)-ACETIC ACID BUTYL ESTER
AldrichCPR
Größe auswählen
5MG
29,10 €
29,10 €
Voraussichtliches Versanddatum31. Mai 2025
Größe auswählen
Ansicht ändern
5MG
29,10 €
About This Item
Lineare Formel:
C24H26O7
CAS-Nummer:
Molekulargewicht:
426.471
MDL-Nummer:
29,10 €
Voraussichtliches Versanddatum31. Mai 2025
Sonstige Hinweise
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)