Merck
MT51302613
Mettler-Toledo SevenGo Duo pH/conductivity SG23
SG68 field kit, InLab® Expert Pro-ISM and InLab® 605-ISM, cable L 5 m
Größe auswählen
1 EA
3.120,00 €
3.120,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
3.120,00 €
About This Item
UNSPSC-Code:
41115603
NACRES:
NB.61
3.120,00 €
Check Cart for Availability
Beschreibung
Calibration pH: Max. 5 points, 7 predefined buffer groups; DO: 1 or 2 points
DO range 0.0 ... 600%, 0.00 ... 99.00 mg/L (+/- 0.5%)
Temperature range °C pH ATC: -5.0 ... 105.0; MTC: -30 ... 130/DO: 0.0 ... 60.0 (+/- 0.2)
Hersteller/Markenname
Mettler-Toledo 51302613
Messbereich mV
-1999.9-1999.9, accuracy: ±0.1
Kabel L
5 m
Akkuleistung
4 AA (included in standard delivery)
pH-Bereich
-2-19.999
Allgemeine Beschreibung
Includes meter, InLab® Expert Pro-ISM and InLab® 605-ISM (both IP67, 5 m cable), rubber holster, calibration sachets, sample bottles, 2-electrode clip and Field Compact Case
SevenGo Duo pro combines maximum measurement accuracy and an excellent range of functions with user-friendly operation and exceptional safety. SevenGo Duo pro meters are essential for special analyses, particularly in regulated sectors in the pharmaceutical, chemical, cosmetics and food & beverage industry. The kit versions are also well suited for outdoor use because both the meter and the sensors are watertight (IP67).
The revolutionary intelligent sensor management system (ISM®) offers the user an even greater level of security. The calibration data is stored on the chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.
Features and Benefits
pH/Ion/Oxygen Meter (IP67) for the Strictest Demands
Portable dual-channel meter for the parallel determination of pH, mV, rel. mV or ion concentrations and dissolved oxygen (DO)
Either linear or segmented 5-point pH calibration with selection from 7 predefined and one user-defined buffer groups
Manual or automatic atmospheric pressure compensation using built-in barometer
Modern data management for 500 GLP data records with date/time, sensor ID and serial number, user and sample ID
Contactless infrared interface for data transmission to a printer or PC
Intuitive, user-friendly operation using menu guidance in 10 languages and backlit display
Rugged watertight housing (IP67)
Revolutionary intelligent sensor management (ISM)
Rechtliche Hinweise
ISM is a registered trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
SevenGo is a trademark of Mettler-Toledo, Inc.
SevenGo Duo pro is a trademark of Mettler-Toledo, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)