MT51302600
Mettler-Toledo SevenGo™ Duo pH/conductivity
SG23-Basic, no electrode
Größe auswählen
1.090,00 €
Größe auswählen
About This Item
1.090,00 €
Empfohlene Produkte
Beschreibung
Calibration Max. 3 points, 4 predefined buffer groups
Conductivity range 0.10 μS/cm ... 500 mS/cm (+/-0.5%)
Resistivity range 0.00 ... 100.00 MOhm*cm
Temperature range °C pH/mV: -5.0 ... 105/LF: -5.0 ... 105.0 (+/- 0.2)
Hersteller/Markenname
Mettler-Toledo 51302600
Messbereich TDS
0.1 mg/L (min)
300 g/L (max)
Messbereich mV
-1999-1999, accuracy: ±1
Akkuleistung
4 AA (included in standard delivery)
pH-Bereich
0-14
Allgemeine Beschreibung
The revolutionary intelligent sensor management system (ISM®) offers the user an even greater level of security. The calibration data is stored on the chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.
Features and Benefits
Routine pH/Conductivity Meter (IP67)
Portable dual-channel meter for the determination of pH, mV, conductivity, TDS (total dissolved solids), salinity and specific resistance
3-point pH calibration with selection from 4 predefined and one user-defined buffer groups
Data memory for 99 GLP data records
Intelligent sensor management (ISM)
Outstanding operation due to high-contrast display with large figures
Reproducible results due to automatic end point detection, automatic buffer detection and temperature compensation
Rugged watertight housing (IP67)
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)