Merck
DWKKC92812G-250
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with KIMCOTE® safety coating and glass stopper
glass flask, flask capacity (250 mL), class A
Größe auswählen
1 EACH
764,00 €
764,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EACH
764,00 €
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
764,00 €
Check Cart for Availability
Materialien
glass flask
Leistungsmerkmale
class A
Verpackung
case of 6 ea
Hersteller/Markenname
Kimble® KC92812G-250
Kolbenkapazität
(250 mL)
Allgemeine Beschreibung
KimCote Class A Heavy Duty Wide Mouth Volumetric Flasks
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Rechtliche Hinweise
KIMBLE is a registered trademark of DWK Life Sciences
KIMCOTE is a registered trademark of DWK LIFE SCIENCES LLC
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)